missing translation for 'onlineSavingsMsg'
Learn More

Bathocuproin sulfonate disodium salt hydrate, 97%

Catalog No. p-7026520
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB22550MD 250 mg
AAB2255003 1 g
AAB2255006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB22550MD Supplier Thermo Scientific Chemicals Supplier No. B22550MD
Only null left

CAS: 52698-84-7 | C26H18N2Na2O6S2 | 564.54 g/mol

Used as a stain for proteins in polyacrylamide gels; for determination of iron and copper; and uric acid by automated proceduresBathocuproin sulfonate disodium salt hydrate is a mixture of isomers suitable for the determination of iron and copper. It is also used as a stain for proteins in polyacrylamide gels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a stain for proteins in polyacrylamide gels; for determination of iron and copper; and uric acid by automated proceduresBathocuproin sulfonate disodium salt hydrate is a mixture of isomers suitable for the determination of iron and copper. It is also used as a stain for proteins in polyacrylamide gels.

Solubility
Miscible with water.

Notes
Incompatible with sulfuric acid, sodium metal, potassium metal and calcium carbide.

Chemical Identifiers

CAS 52698-84-7
Molecular Formula C26H18N2Na2O6S2
Molecular Weight (g/mol) 564.54
MDL Number MFCD00149974
InChI Key RNGKZLRAVYPLJC-UHFFFAOYSA-L
Synonym disodium bathocuproinedisulfonate, sodium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate, bathocuproinedisulfonic acid sodium salt, 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline disulfonate, 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline disulfonate disodium salt, dipotassium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate, disodium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate
PubChem CID 15678335
SMILES [Na+].[Na+].CC1=NC2=C(C=CC3=C2N=C(C)C(=C3C2=CC=CC=C2)S([O-])(=O)=O)C(C2=CC=CC=C2)=C1S([O-])(=O)=O

Specifications

Melting Point >300°C
Quantity 250 mg
Beilstein 9033637
Solubility Information Miscible with water.
IUPAC Name disodium;2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate
Formula Weight 564.55 (Anhydrous)
Percent Purity 97%
Chemical Name or Material Bathocuproin sulfonate disodium salt hydrate
EINECSNumber 258-111-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only