Learn More
Chemglass Life Sciences Oil Bath Fluid, 1/2 Gallon DFS Item
Supplier:  Chemglass Inc CG110031
Oil Bath Fluid, 1/2 Gallon. High temperature silicone bath fluid for use with CG-1100 series oils bath dishes.Viscosity (@25 Deg C) 100cst Operating Temperatures, Deg C -40 to 315 Flashpoint Deg C (Open Cup) >288 Pour Point (Deg C) -65 Coefficient of Expansion 0.000095 cc/cc/DegC Specific Heat, Cal/gm/Deg C 34 Thermal Conductivity, gm- 0.00026 cal/s-cm2/Deg C
Catalog No. CG110031
Only null left
Explore available promotions