Learn More
Enzo Life Sciences Bassoon monoclonal antibody (SAP7F407) (50 g)
Supplier: Enzo ENZABS6660050
Clone: SAP7F407. Host: Mouse. Isotype: IgG2a. Immunogen: Recombinant rat Bassoon. UniProt: O88778. Source: Purified from hybridoma tissue culture supernatant. Species Reactivity: Human, Mouse, Rat, Dog, Bovine. Applications: ICC, IF, IHC, IP, WB. Recommended dilutions: Immunocytochemistry (1:400). Immunoprecipitation (2.5g). Western Blot (1:500 - 1:1,000). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.