Learn More
Elma™ Baskets for Elmasonic E Plus Ultrasonic Cleaners
Holds parts while immersing, removing from the cleaner solution. Tovatech Baskets for Elmasonic E Plus Ultrasonic Cleaners protect the parts from damage caused by the cleaners.
Supplier: Elma™ 1004178
Description
- Solid-sides with mesh bottom
- Plastic coated handles for insulation
- Allows parts to be removed from hot cleaning solution
- Stainless steel construction
Specifications
| Basket | |
| Solid-Side Basket with Mesh Bottoms | |
| 7.62 cm | |
| Stainless Steel |
| Ultrasonic Cleaner, Elmasonic E Plus 100 H | |
| Hold Parts When Immersing and Removing Them From The Solution in Ultrasonic Cleaners | |
| 25.4 cm | |
| 19.81 cm |