missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Basic Vortex Mixers

Catalog No. 88882011
Change view
Click to view available options
Plug Type:
US Plug
Certifications/Compliance:
cCSAus
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Type Certifications/Compliance
88882011 US Plug cCSAus
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 88882011 Supplier Thermo Scientific™ Supplier No. 88882011
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Standard package includes the Rubber platform 88882120 x 1

The Thermo Scientific™ Basic Vortex Mixer is ideal for quick, uniform mixing of samples with precise speed control.

The Thermo Scientific™ Basic Vortex Mixer provides comprehensive mixing capabilities for enzymatic and RIA assays, general test tube mixing, biochemical assays, viral dilution preparation, precipitated assays, cell suspension vortexing and tissue sample mixing.

  • Digital Speed display
  • Continuous and Touch modes
  • Speed range: 200 to 3000rpm
  • Speed Accuracy: ± 5% rpm
  • Convenient Speed adjustments by knob control
  • Stable for long-term operations
  • Multiple accessories for different vessels

Order Info

Please order various accessories as the adapter for multiple applications

Specifications

Orbit 4 mm
Load Bearing Capacity (Metric) 0.5 kg
Speed Range 200 to 3000 rpm
Electrical Requirements 100/240 V
Voltage 120 V
Holds Tubes
Product Type Basic Vortex Mixer
Certifications/Compliance cCSAus
Plug Type US Plug
Frequency 50/60 Hz
Material ABS housing
Dimensions (L x W x H) 290 x 270 x 240 mm (11.4 x 10.7 x 9.4 in.)
Length (English) 11.4 in.
Length (Metric) 290 mm
Shipping Weight (English) 14.99 lb.
Shipping Weight (Metric) 6.8 kg
Weight (English) 14.33 lb.
Weight (Metric) 6.5 kg
Model Basic Vortex Mixer
Display Digital
For Use With (Equipment) Accessories: 88882120, 88882121, 88882122, 88882123, 88882124, 88882125, 88882126, 88882127
Includes Rubber platform 88882120
IP Rating IP32
Motor Type DC motor
No. of Positions 19
Operating Modes Touch, Continous
Warranty and Service Offering 2 Years
Show More Show Less