Learn More
Eisco™ Basic Spectrometer
An economical spectrometer for use in elementary schools to measure the angle of prisms using the law of reflection, demonstrate optics phenomenon with a diffraction grating or measure the wavelengths of monochromatic light sources.
Supplier:  Eisco™ PH0618A
Catalog No. S23680
Only null left
Explore available promotions
Includes:
Adjustable collimator tube, telescope, and prism stand
Description
Description
  • Built from sturdy metal
  • Students obtain reliable results when they work with this instrument
  • Supplied in strong cardboard box
Specifications
Specifications
Basic Spectrometer
Metal
Measure the angle of prisms using the law of reflection, demonstrate optics phenomenon with a diffraction grating or measure the wavelengths of monochromatic light sources
Adjustable collimator tube, telescope, and prism stand
Product Suggestions
Product Suggestions