Learn More
United Scientific Supplies Basic Solar Kit
Catalog No. S27844
Change view
Click to view available options
Product Type:
Kit, Solar
Catalog No. Product Type
S27844 Kit, Solar
1 options
Catalog No. S27844 Supplier United Scientific Supplies Supplier No. SLRKITB
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A convenient introduction to the conversion of solar light energy into electrical energy
  • Convenient kit provides everything needed to convert solar-generated electrical energy into light, sound, or motion
  • Observe the correlation between angle of incidence and energy output
Specifications
Product Type Kit, Solar
For Use With (Application) Converting Solar-Generated Electrical Energy Into Light, Sound, or Motion
Includes Solar Panel (1), Red LED (1), Buzzer (1), Motor with Fan (1), Mirror (1), and Activity Guide (1)