missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Basic Security Module, FlexMAP™ 3D™ System

Catalog No. FM3D0BS
Change view
Click to view available options
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FM3D0BS 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FM3D0BS Supplier Invitrogen™ Supplier No. FM3D0BS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The basic security module provides security by allowing an administrator to set up user access levels.

The basic security module provides security by allowing an administrator to set up user access levels. This module is designed for the FlexMAP™ 3D™ system.

Specifications

For Use With (Equipment) FlexMAP™ 3 D™ System
Product Line Luminex
Quantity 1 each
Content And Storage Store at room temperature
Shipping Condition Room Temperature
Software Security
How long does it take to acquire ProcartaPlex assay data on the different Luminex instruments?

With everything working smoothly, here is the approximate read time for one plate on different Luminex instruments:

- FLEXMAP 3D Systems and INTELLIFLEX Systems will read 96 well plates in ~20 min, and 384 well plates in ~75 min.
- Luminex 100/200 Systems will read 96 well plates in ~40 min.
- MAGPIX System will read 96 well plates in ~50-60 min. Please note that these are estimates for the read times of the plates only, and do not include time for calibration or set-up of the instrument.