Learn More
Eisco™ Basic Optical Bench
Catalog No. S23750
Change view
Click to view available options
Length (Metric):
1 m
Catalog No. Length (Metric)
S23750 1 m
1 options
Catalog No. S23750 Supplier Eisco™ Supplier No. PH0616OPT
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
(1) Meter rule rod, (4) Riders , (1) Slit holder, (1) Candle Holder, (1) Object Needle, (1) Object Screen, (2) Lens Holders
Upgrade from basic optics meter rule bench for introductory optics investigations.
  • Comprises of square section aluminum tube
  • Tube graduated on one side allowing easy reading when riders move on bench
Specifications
Product Type Basic Optical Bench
Description 4 riders
For Use With (Application) introductory optics investigations
Includes (1) Meter rule rod, (4) Riders , (1) Slit holder, (1) Candle Holder, (1) Object Needle, (1) Object Screen, (2) Lens Holders
Length (Metric) 1 m