missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Basic Fuchsin, 70+%, pure

Catalog No. AC401691000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC401691000 100 g Glass bottle
AC401695000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC401691000 Supplier Thermo Scientific Chemicals Supplier No. 401691000

CAS: 632-99-5 | C20H20ClN3 | 337.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 632-99-5
Molecular Formula C20H20ClN3
Molecular Weight (g/mol) 337.85
MDL Number MFCD00012569
InChI Key AXDJCCTWPBKUKL-UHFFFAOYSA-N
Synonym Basic Violet 14, hydrochloride, C.I. 42510, Rosaniline chloride
PubChem CID 12447
IUPAC Name 4-[(4-aminophenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride
SMILES [H+].[Cl-].CC1=CC(C=CC1=N)=C(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1

Specifications

Melting Point 250.0°C
Absorption Ratio 1.16 to 1.35
Dye Content 70% min.
Beilstein 13, 765
Merck Index 15, 5715
Solubility Information Solubility in water: 4g/L (25°C). Other solubilities: soluble in alcohol and acids, practically insoluble in ether, soluble 30g/L in ethanol (25°C)
Infrared Spectrum Authentic
Lambda Max 547 to 552nm
Formula Weight 337.84
Percent Purity ≥70%
Grade Pure
Packaging Glass bottle
Density 200g/mL
Color Green
Quantity 100 g
Flash Point 200°C
Physical Form Glistening Crystalline Powder
Chemical Name or Material Basic Fuchsin, Pure
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Suspected of causing cancer. Harmful if swallowed.
Precautionary Statement GHS P Statement
Use personal protective equipment as required. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only