Learn More
Science First™ Basic Electric Motor Kit
Catalog No. S43597
Change view
Click to view available options
Voltage:
1.5 VDC
Catalog No. Voltage
S43597 1.5 VDC
1 options
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Catalog No. S43597 Supplier Science First™ Supplier No. 10 135 P12
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Includes
Coil of copper wire, plastic base, field poles, armature core, brushes, fasteners and instructions with eight experiments
Requires
One AA battery (S43921)
Build a real working motor for classroom exercises, science fairs and more.
  • Build a working DC motor using detailed instructions featuring computer-generated assembly diagrams
  • Easy assembly: wind the armature and field coil, build the commutator, install brushes into holes in the base and slip a battery into battery clips
Order Info
Grades 5+.
Specifications
For Use With (Application) Classroom exercises, science fairs and more, for basic training in electricity
Voltage 1.5 VDC
Includes Coil of Copper wire, Plastic base, field pole, armature core, brushes, fasteners and instruction with eight experiment