missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Basic Blue 24, TCI America™
Specifications
Specifications
| Color | Black/Yellow |
| Quantity | 5 g |
| Physical Form | Crystal/Powder |
| Chemical Name or Material | Basic Blue 24 |
Safety and Handling
| TSCA | No |