Learn More
Pyramex Safety Products Baseball Bump Cap
Catalog No. 19555098
Change view
Click to view available options
Color:
Hi-vis Orange
Navy Blue
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19555098 Hi-vis Orange
19555099 Navy Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19555098 Supplier Pyramex Safety Products Supplier No. HP50041
Only null left
Pyramex Safety™ Baseball Bump Cap features reflective piping for extra visibility.
  • Soft inner foam cushion for comfortable wear
  • Size can be adjusted with a hook and loop strap to give a quick, secure fit.
  • Tough polyethylene vented shell is removable and the outer cap is washable.
  • Ideal for food processing and other industries that don’t require full hard hat protection.
  • Holes in the liner match up with two mesh panels on the cap to provide additional ventilation for coolness and comfort.
Specifications
Color Hi-vis Orange
Material Polyethylene
Size Adjustable