missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Base Retort Stand

Rectangular in shape.

Supplier:  Eisco™ CH0652C

 View more versions of this product

Catalog No. S02612


Only null left

Description

Description

  • Pressed steel base
  • With hole 10 x1.5mm metric thread in the center of one short side
  • Finished in blue/grey colour
Specifications

Chemical Identifiers

Steel
Rectangular
Blue/Gray
Base Retort Stand
0.39 x 0.05 in.
Each

Specifications

Steel
1.1 in.
25 cm
16 cm
Rectangular
Blue/Gray
5.5 lb.
3 cm
9.8 in.
6.2 in.
Base Retort Stand
0.39 x 0.05 in.
Each
2.5 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions