missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Basal Cell Culture Liquid Media - DMEM and Ham's F-12, 50/50 Mix

Catalog No. p-2410144
Change view
Click to view available options
Quantity:
6 x 500 mL
6 x 1 L
With Additives:
Glutamine
Glutamine, HEPES
None
Without Additives:
No Glutamine
No Phenol Red
None
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity With Additives Without Additives
MT15090CM 6 x 1 L None No Glutamine
MT10092CV 6 x 500 mL Glutamine, HEPES None
MT16405CV 6 x 500 mL Glutamine No Phenol Red
MT10090CV 6 x 500 mL Glutamine None
MT10092CM 6 x 1 L Glutamine, HEPES None
MT15090CV 6 x 500 mL None No Glutamine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MT15090CM Supplier Corning™ Supplier No. 15090CM
Only null left

Used for culture of epithelial, endothelial, and granulosa cell types under low serum conditions. Corning™ Basal Cell Culture Liquid Media - DMEM and Ham's F-12, 50/50 Mix has low levels of endotoxin, and absence of contamination.

  • Modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME
  • Fifty-fifty mixture of DMEM and F-12, originally formulated for rat neuroblastoma cells and MDCK cells
  • Serum-free growth may be possible with appropriate protein supplements
  • Ham's F-12 media is based on Ham's F-10 medium with increased concentrations of choline, inositol, putrescine, and several amino acids
  • Formulation of DMEM/Ham's F-12 contains L-glutamine
  • Sterile

Specifications

Concentration 1 X
Form Liquid
Product Type DMEM and Ham's F-12, 50/50 Mix
Sterility Sterile
With Additives None
Without Additives No Glutamine
For Use With (Application) To assess growth promotion and cytotoxicity characteristics
Quantity 6 x 1 L
Endotoxin Level <0.25EU/mL
pH 7 to 7.4
Content And Storage 2° to 8°C
Show More Show Less

IVD Product