Learn More
Thermo Scientific™ Barnstead™ Mega-Pure™ Accessories
Catalog No. p-4531428
Change view
Click to view available options
Description:
Dual Solenoid, MP-1A (240V), MP-3, MP-6A, and MP-11A
High Capacity Cartridge
High Purity Cartridge
Organic Removal Cartridge
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
0903593 Dual Solenoid, MP-1A (240V), MP-3, MP-6A, and MP-11A
0903578 High Purity Cartridge
0903568 High Capacity Cartridge
0903561 Organic Removal Cartridge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 0903593 Supplier Thermo Scientific™ Supplier No. 440236
Select from an array of Thermo Scientific™ Barnstead™ Mega-Pure™ Accessories for your Mega-Pure™ Glass Still, including storage bottles and deionization cartridges.
Select from an array of Thermo Scientific™ Barnstead™ Mega-Pure™ Accessories for your Mega-Pure Glass Still. Accessories include dual solenoid, deionization cartridges, storage solutions, single cartridge deionizer, dual cartridge deionizer, deionizer accessories and cartridges, storage bottles and automatic collection system.
Specifications
Description Dual Solenoid, MP-1A (240V), MP-3, MP-6A, and MP-11A
Type Dual Solenoid
For Use With (Equipment) MegaPure Glass Still