missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Barnstead™ Hose Nipple Cartridges

Catalog No. 0903520
Change view
Click to view available options
Description:
2/3 Size High Capacity
2/3 Size Mixed-Bed and Organic Removal
Full Size High Capacity
Full Size Organic Removal
Full Size Oxygen Removal
Full Size Pretreatment and Scale Eliminator
Full Size Ultrapure
Full Size Ultrapure with Organic Removal
Hose Nipple Cartridge Holder
Hose nipple cartridge, Full Size Cation Removal
Resin Type:
Activated Carbon
Cation Exchange
Mixed-bed and Strong Cation Exchange, Activated Charcoal
Mixed-bed Ion Exchange with Activated Carbon
Strong Anion Exchange
Two Bed Ion Exchange
Two Bed Ion Exchange with Strong Cation Exchange
Ultrapure Ion Exchange
Ultrapure Mixed Bed Ion Exchange
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Resin Type
0903520 Full Size High Capacity Two Bed Ion Exchange
0903525 Full Size Ultrapure Ultrapure Mixed Bed Ion Exchange
0903411 Full Size Oxygen Removal Strong Anion Exchange
0903522 Full Size Organic Removal Activated Carbon
0903412 Hose nipple cartridge, Full Size Cation Removal Cation Exchange
0903414 Full Size Ultrapure Ultrapure Ion Exchange
0903532 Full Size Pretreatment and Scale Eliminator Mixed-bed and Strong Cation Exchange, Activated Charcoal
0903530 Full Size Ultrapure with Organic Removal Mixed-bed Ion Exchange with Activated Carbon
0903528 2/3 Size High Capacity Two Bed Ion Exchange with Strong Cation Exchange
09050272 2/3 Size Mixed-Bed and Organic Removal Mixed-bed Ion Exchange with Activated Carbon
0903550 Hose Nipple Cartridge Holder -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 0903520 Supplier Thermo Scientific™ Supplier No. D 8901

Purify water up to 75 L/hr using these hose nipple cartridges, ideal for routine lab work including pretreatment.

Purify water volumes up to 75L/hr using cost-efficient Thermo Scientific™ Barnstead™ Hose Nipple Cartridges. Ideal for softening, deionization, and organic and chlorine removal applications, these cartridges are for use with the simple cartridge holder that easily mounts to the wall. Designed for non-pressure applications, these cartridges include 3/8 in. barb hose connections on each end. They provide the ability to see when resin is exhausted in specific cartridges by a color change indicator. Constructed of 100% virgin polypropylene cartridge, the system is customizable and an ideal option for routine laboratory work.

  • Ideal for small volume applications
  • Same superior quality resins used in B-Pure cartridges
  • Constructed of 100% virgin polypropylene cartridge
  • Includes 3/8 in. barb hose connections on each end
  • D8822, D8950, and D8951 cartridges include tapered straight nipple for 3/8 in. ID tubing
  • Color change indicator that identifies when resin is exhausted in specific cartridges

Specifications

Description Full Size High Capacity
For Use With (Equipment) Barnstead Water Purification System
Particle Retention 1650 grains
Type Cartridge, Hose Nipple
Flow Rate Up to 75 L/hr.
For Use With (Application) Two Bed Ion Exchange
Includes Color Indicator
Resin Type Two Bed Ion Exchange