Learn More
Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables
Catalog No. BSSLMX13
Change view
Click to view available options
Description:
Cartridge Kit
Replacement Ultraviolet Lamp
Start-up Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
BSSLMX13 Replacement Ultraviolet Lamp
1393581 Start-up Kit
1393582 Cartridge Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BSSLMX13 Supplier Thermo Scientific™ Supplier No. LMX13
Customize your water purification system with the Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables that best suit your application.
Customize your water purification system with the Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables that best suit your application. Cartridge kit includes pre-filter, ultrapure mixed bed cartridge and Easypure high purity/low TOC cartridge. Other consumables include 0.2 micron final filter, ultraviolet lamp, Thermo Scientific™ Ventgard™ reservoir cap, reverse osmosis membrane, and set of three empty cartridges for cleaning.
Specifications
Description Replacement Ultraviolet Lamp
For Use With (Equipment) Barnstead Easypure RoDi
Type UV Lamp