Learn More
Thermo Scientific™ Barnstead™ E-Pure™ Optional Accessories
Catalog No. 0903534
Change view
Click to view available options
Type:
Remote Dispenser
Voltage:
0/1 V
Catalog No. Type Voltage
0903534 Remote Dispenser 0/1 V
1 options
Catalog No. 0903534 Supplier Thermo Scientific™ Supplier No. D8952
Optimize your ultrapure water purification system with Thermo Scientific™ Barnstead™ E-Pure™ Optional Accessories, including a remote dispenser.
Optimize your ultrapure water purification system with Thermo Scientific™ Barnstead™ E-Pure™ Optional Accessories. Increase flexibility in your lab with the remote dispenser, which delivers water up to 8' (2.4m) from the system and ships with a mounting bracket. Gain protection with the low pressure pump protector which alerts the E-Pure of an inadequate feed water condition in a pressurized feed line. The low water level pump protector alerts the system of an inadequate feed water condition when fed from a reservoir.
Specifications
For Use With (Equipment) Barnstead E-Pure
Type Remote Dispenser
Voltage 0/1 V