Learn More
Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits
Description
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits. These cartridge kits are classified based on type of feed water used; choose the appropriate kit according to your feed water and need for organic free water.
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits. These cartridge kits are classified based on type of feed water used; choose the appropriate kit according to your feed water and need for organic free water.
Specifications
Specifications
| Description | Cartridge Kit for E-Pure 4-holder; Pretreated feed, Organic free |
| Feedwater Source | Pretreated |
| For Use With (Equipment) | E-Pure 4-holder, Organic free |
| Type | Cartridge Kit |