Learn More
Thermo Scientific™ Barnstead™ Classic Stills Replacement Consumables
Catalog No. 09050116
Change view
Click to view available options
Type:
Replacement Filter
Replacement UV Lamp
For Use With (Equipment):
Barnstead Classic Stills
Barnstead Classic Stills Reservoirs, H4005
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
09050116 Replacement Filter Barnstead Classic Stills
0902845 Replacement UV Lamp Barnstead Classic Stills Reservoirs, H4005
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09050116 Supplier Thermo Scientific™ Supplier No. H3120
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Maintain the performance of your Thermo Scientific™ Barnstead™ Classic Still with replacement consumables such as UV lamps and air filters.
Maintain the performance of your still with Thermo Scientific™ Barnstead™ Classic Stills Replacement Consumables, including UV lamp and Thermo Scientific™ Ventgard™ filter element (the consumable portion of the Ventgard filter assembly). Prevent air from entering the reservoir via the condenser atmospheric vent and protect stored water against airborne particulates, organics and CO2 with the Ventgard air filter.
Maintain the performance of your still with Thermo Scientific™ Barnstead™ Classic Stills Replacement Consumables, including UV lamp and Thermo Scientific™ Ventgard™ filter element (the consumable portion of the Ventgard filter assembly). Prevent air from entering the reservoir via the condenser atmospheric vent and protect stored water against airborne particulates, organics and CO2 with the Ventgard air filter. 
Specifications
Type Replacement Filter
For Use With (Equipment) Barnstead Classic Stills
Features Protects stored water against airborne particulates and organics.