missing translation for 'onlineSavingsMsg'
Learn More

Barium titanium oxide, high surface area

Catalog No. p-7051601
Change view
Click to view available options
Quantity:
100 g
25 g
Catalog No. Quantity
AA4163314 25 g
AA4163322 100 g
2 options

Catalog No. AA4163314

Supplier: Thermo Scientific Chemicals 04163314

Only null left

CAS: 12047-27-7 | BaO3Ti | 233.19 g/mol

Barium titanium oxide is a dielectric ceramic widely used in multi-layer ceramic capacitors (MLCCs), embedded decoupling capacitors (EDC) and ceramic filters. It is a piezoelectric and ferroelectric component used in microphones and transducers. It is also useful for thermistors and self-regulating electric heating systems. It is doped with small amounts of metals like scandium, yttrium, neodymium, samarium to become a semiconductor. It is used in some types of uncooled sensors for thermal cameras and in electric vehicles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12047-27-7
Molecular Formula BaO3Ti
Molecular Weight (g/mol) 233.19
MDL Number MFCD00003447
InChI Key WNKMTAQXMLAYHX-UHFFFAOYSA-N
Synonym Barium titanate
IUPAC Name barium(2+) oxotitaniumbis(olate)
SMILES [Ba++].[O-][Ti]([O-])=O

Specifications

Melting Point 1620°C
Color White to Gray
Physical Form Powder
Quantity 25 g
Merck Index 14,1000
Solubility Information Slightly soluble in dilute mineral acids. Soluble in concentrated hydrofluoric acid. Insoluble in water.
Formula Weight 233.19
Percent Purity ≥99%
Odor Odorless
Density 5.85 g/mL
Assay (metals basis)
Chemical Name or Material Barium titanium oxide, High surface area
Show More Show Less
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
EINECSNumber 234-975-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only