missing translation for 'onlineSavingsMsg'
Learn More

Barium titanium oxide, 99%

Catalog No. p-7048982
Change view
Click to view available options
Quantity:
50 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1234818 50 g
AA1234836 500 g
AA12348A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1234818 Supplier Thermo Scientific Chemicals Supplier No. 01234818
Only null left

Ferroelectric ceramics, single crystals, storage devices, dielectric amplifiers | CAS: 12047-27-7 | BaO3Ti | 233.19 g/mol

Barium titanium oxide is a dielectric ceramic used for capacitors. It is a piezoelectric material for microphones and other transducers. Finds applications in nonlinear optics and photorefractive applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12047-27-7
Molecular Formula BaO3Ti
Molecular Weight (g/mol) 233.19
MDL Number MFCD00003447
InChI Key WNKMTAQXMLAYHX-UHFFFAOYSA-N
Synonym Barium titanate
IUPAC Name barium(2+) oxotitaniumbis(olate)
SMILES [Ba++].[O-][Ti]([O-])=O

Specifications

Melting Point 1620°C
Physical Form Powder
Quantity 50 g
Merck Index 14,1000
Solubility Information Insoluble in water, alkalies; Slightly soluble in dilute acids; Soluble in concentrated acids
Formula Weight 233.19
Percent Purity 99%
Odor Odorless
Density 5.85 g/mL
Assay (metals basis)
Chemical Name or Material Barium titanium oxide
Show More Show Less
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
EINECSNumber 234-975-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only