missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium titanium oxide, 99.95% (metals basis)
CAS: 12047-27-7 | BaO3Ti | 233.19 g/mol
$163.64 - $661.99
Chemical Identifiers
| CAS | 12047-27-7 |
|---|---|
| Molecular Formula | BaO3Ti |
| Molecular Weight (g/mol) | 233.19 |
| MDL Number | MFCD00003447 |
| InChI Key | WNKMTAQXMLAYHX-UHFFFAOYSA-N |
| Synonym | Barium titanate |
| IUPAC Name | barium(2+) oxotitaniumbis(olate) |
| SMILES | [Ba++].[O-][Ti]([O-])=O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 12047-27-7 | |
| 233.19 | |
| WNKMTAQXMLAYHX-UHFFFAOYSA-N | |
| barium(2+) oxotitaniumbis(olate) |
| BaO3Ti | |
| MFCD00003447 | |
| Barium titanate | |
| [Ba++].[O-][Ti]([O-])=O |
Specifications
| 1620°C | |
| Pieces | |
| BaO3Ti | |
| 14,1000 | |
| WNKMTAQXMLAYHX-UHFFFAOYSA-N | |
| barium(2+) oxotitaniumbis(olate) | |
| 233.19 | |
| Odorless | |
| (metals basis) |
| 12047-27-7 | |
| 5 g | |
| MFCD00003447 | |
| Barium titanate | |
| [Ba++].[O-][Ti]([O-])=O | |
| 233.19 | |
| 99.95% | |
| Barium titanium oxide |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
EINECSNumber : 234-975-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only