missing translation for 'onlineSavingsMsg'
Learn More

Barium titanate(IV), 99%

Catalog No. p-222683
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC196865000 500 g Plastic bottle
AC196860025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC196865000 Supplier Thermo Scientific Chemicals Supplier No. 196865000
Only null left

CAS: 12047-27-7 | BaO3Ti | 233.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12047-27-7
Molecular Formula BaO3Ti
Molecular Weight (g/mol) 233.19
MDL Number MFCD00003447
InChI Key WNKMTAQXMLAYHX-UHFFFAOYSA-N
IUPAC Name barium(2+) oxotitaniumbis(olate)
SMILES [Ba++].[O-][Ti]([O-])=O

Specifications

Melting Point 1625.0°C
Color White
CAS Min % 98.5
CAS Max % 100.0
Physical Form Powder
Quantity 500 g
Assay Percent Range 0.988 to 1.000 (BaO+ SrO)/TiO2 Mole Ratio
Linear Formula BaTiO3
Merck Index 15, 996
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in alkalies, slightly soluble in diluted acids, soluble in concentrated h2so4 and hf
Formula Weight 233.24
Percent Purity 99%
Loss on Ignition 0.5% max.
Packaging Plastic bottle
Specific Gravity 6.08
Density 6.0800g/mL
Chemical Name or Material Barium titanate(IV), 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
EINECSNumber 234-975-0
RTECSNumber XR1437333
TSCA TSCA

RUO – Research Use Only