missing translation for 'onlineSavingsMsg'
Learn More

Barium Sulfide, Black, Granular, 80%, Spectrum™ Chemical

$315.91 - $2745.65

Chemical Identifiers

CAS 8011-62-9
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18609031
View Documents
Spectrum Chemical Mfg Cor
B1060125GM
125 g Poly Bottle
Each for $315.91
Only null left
 
18609032
View Documents
Spectrum Chemical Mfg Cor
B1060500GM
500 g Poly Bottle
Each for $774.50
Only null left
 
18609033
View Documents
Spectrum Chemical Mfg Cor
B106025KG
2.5 kg Poly Bottle
Each for $2,745.65
Only null left
 
Description

Description

Specifications

Chemical Identifiers

8011-62-9

Specifications

8011-62-9
125 g
Reagent
1
80%
Poly Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.