missing translation for 'onlineSavingsMsg'
Learn More

Barium Sulfate, USP, 97.5-100.5%, Spectrum™ Chemical

Catalog No. 18600848
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
25 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600848 100 g Poly Bottle
18600849 500 g Poly Bottle
18600850 2.5 kg Poly Bottle
18600852 25 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600848 Supplier Spectrum Chemical Mfg Cor Supplier No. BA125100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

BA125, 7727-43-7, BaSO4

Spectrum™ Chemical Barium Sulfate, USP is a contrast medium for CT scans or fluoroscopic studies of the gastrointestinal tract. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7727-43-7
Molecular Formula BaO4S
Molecular Weight (g/mol) 233.38
MDL Number MFCD00003455
InChI Key TZCXTZWJZNENPQ-UHFFFAOYSA-L
IUPAC Name barium(2+) sulfate
SMILES [Ba++].[O-]S([O-])(=O)=O

Specifications

pH 3.5-10.0
CAS Max % 100%
Physical Form Crystalline Powder or Needles
Quantity 100 g
Percent Purity 97.5 to 100.5%
Grade USP
Packaging Poly Bottle
Chemical Name or Material Barium Sulfate