missing translation for 'onlineSavingsMsg'
Learn More

Barium Sulfate (Powder/USP), Fisher Chemical™

Catalog No. p-31163
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
B75500 500 g Poly Bottle
B75212 2.5 kg Poly Bottle
B7512 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B75500 Supplier Thermo Fisher Scientific Supplier No. FLB75 500

Fine white powder, free from grittiness

Chemical Identifiers

CAS 7727-43-7
Molecular Formula BaO4S
Molecular Weight (g/mol) 233.38
MDL Number MFCD00003455
InChI Key TZCXTZWJZNENPQ-UHFFFAOYSA-L
Synonym barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast
PubChem CID 24414
IUPAC Name barium(2+) sulfate
SMILES [Ba++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1580°C
Color Yellow
Physical Form Solid
Quantity 500 g
Assay Percent Range 97.5 to 100.5 %
Formula Weight 233.39
Percent Purity 97.5 to 100.5%
Grade USP
Identification Pass Test
Packaging Poly Bottle
Chemical Name or Material Barium Sulfate
Show More Show Less