missing translation for 'onlineSavingsMsg'
Learn More

Barium peroxide, anhydrous, 84% min

Catalog No. p-4781808
Change view
Click to view available options
Quantity:
1 kg
250 g
5 kg
50 g
Catalog No. Quantity
AA1400818 50 g
AA1400830 250 g
AA14008A1 1 kg
AA14008A7 5 kg
4 options

Catalog No. AA1400818

Supplier: Thermo Scientific Chemicals 01400818

Only null left

CAS: 1304-29-6 | BaO2 | 169.325 g/mol

Barium peroxide is a specialty product for proteomics research. It is a strong oxidizing agent. It is also used for bleaching in a variety of commercial applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1304-29-6
Molecular Formula BaO2
Molecular Weight (g/mol) 169.325
MDL Number MFCD00003454
InChI Key ZJRXSAYFZMGQFP-UHFFFAOYSA-N
Synonym barium peroxide, barium dioxide, barium binoxide, barium superoxide, barium oxide, per, bario perossido di, bariumperoxid german, bariumperoxyde dutch, dioxyde de baryum french, barium peroxide ba o2
PubChem CID 14773
IUPAC Name barium(2+);peroxide
SMILES [O-][O-].[Ba+2]

Specifications

Melting Point 450°C (decomposition)
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 84%
Solubility Information Insoluble in water
Formula Weight 169.34
Odor Odorless
Sensitivity Moisture sensitive
Density 4.96 g/cm3
Chemical Name or Material Barium peroxide
Show More Show Less
Hazard Category H272-H302+H332
Hazard Statement Keep away from food, drink and animal feeding stuffs. Avoid contact with skin and eyes. Take off immediately all contaminated clothing.
Precautionary Statement P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P312-P330-P370+P378q-P501c
DOTInformation DOT Class: 5.1, Packaging Group: II
EINECSNumber 215-128-4
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only