missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium perchlorate solution, Solstice
Description
Solstice's high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
| Physical Form | Liquid |
| Quantity | 1 L |
| Flash Point | 21°C (69.8°F) |
| Linear Formula | Ba(ClO4)2 |
| UN Number | UN1219 |
| Formula Weight | 336.23g/mol |
| Grade | Not Available |
| Chemical Name or Material | Barium perchlorate solution |
Safety and Handling
| Hazard Statement | P210-P305 + P351 + P338-P370 + P378-P403 + P235 |