missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium perchlorate, anhydrous
Description
Barium perchlorate, anhydrous is used to make explosives. It is also used as desiccant, ribonuclease determination.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless |
| Physical Form | Powder |
| Quantity | 100 g |
| Solubility Information | Soluble in water. |
| Formula Weight | 336.24 |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Density | 3.20 g/cm3 |
| Chemical Name or Material | Barium perchlorate |
Safety and Handling
| missing translation for 'healthHazard1' | H271-H302+H332 |
| missing translation for 'healthHazard3' | P210-P220-P221-P261-P264b-P270-P271-P280-P283-P301+P312-P304+P340-P306+P360-P312-P330-P370+P378q-P371+P380+P375-P501c |
| DOTInformation | DOT Class: 5.1, Packaging Group: II |
| EINECSNumber | 236-710-4 |
| RTECSNumber | SC7550000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only