missing translation for 'onlineSavingsMsg'
Learn More

Barium perchlorate, anhydrous

Catalog No. aa1165736
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1165736 500 g
AA1165722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1165736 Supplier Thermo Scientific Chemicals Supplier No. 01165736
Only null left

CAS: 13465-95-7 | BaCl2O8 | 336.219 g/mol

Barium perchlorate, anhydrous is used to make explosives. It is also used as desiccant, ribonuclease determination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13465-95-7
Molecular Formula BaCl2O8
Molecular Weight (g/mol) 336.219
MDL Number MFCD00003440
InChI Key OOULUYZFLXDWDQ-UHFFFAOYSA-L
Synonym barium perchlorate, barium diperchlorate, perchloric acid, barium salt, unii-ey48pa0c98, barium 2+ diperchlorate, barium 2+ ion diperchlorate, barium perchlorate, anhydrous, perchloric acid, bariumsalt 2:1, perchloric acid, barium salt 2:1, acmc-20alqv
PubChem CID 61623
IUPAC Name barium(2+);diperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ba+2]

Specifications

Color Colorless
Physical Form Powder
Quantity 500 g
Solubility Information Soluble in water.
Formula Weight 336.24
Odor Odorless
Sensitivity Hygroscopic
Density 3.2
Chemical Name or Material Barium perchlorate
Hazard Category H271-H302+H332
Precautionary Statement P210-P220-P221-P261-P264b-P270-P271-P280-P283-P301+P312-P304+P340-P306+P360-P312-P330-P370+P378q-P371+P380+P375-P501c
DOTInformation DOT Class: 5.1, Packaging Group: II
EINECSNumber 236-710-4
RTECSNumber SC7550000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only