missing translation for 'onlineSavingsMsg'
Learn More

Barium perchlorate, 99%, extra pure, anhydrous

Catalog No. p-222679
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC221271000 100 g Glass bottle
AC221275000 500 g Glass bottle
2 options

Catalog No. AC221271000

Supplier: Thermo Scientific Chemicals 221271000

Only null left

CAS: 13465-95-7 | BaCl2O8 | 336.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent

Chemical Identifiers

CAS 13465-95-7
Molecular Formula BaCl2O8
Molecular Weight (g/mol) 336.23
MDL Number MFCD00003440
InChI Key OOULUYZFLXDWDQ-UHFFFAOYSA-L
Synonym barium perchlorate, barium diperchlorate, perchloric acid, barium salt, unii-ey48pa0c98, barium 2+ diperchlorate, barium 2+ ion diperchlorate, barium perchlorate, anhydrous, perchloric acid, bariumsalt 2:1, perchloric acid, barium salt 2:1, acmc-20alqv
PubChem CID 61623
IUPAC Name barium(2+);diperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ba+2]

Specifications

Melting Point 505.0°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 98.5% min. (Complexometry)
Linear Formula Ba(ClO4)2
Merck Index 15, 983
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, slightly soluble in ethanol, ethyl acetate and, acetone
Formula Weight 336.23
Percent Purity 99%
Grade Extra Pure
Packaging Glass bottle
Specific Gravity 3.2
Density 3.2000g/mL
Chemical Name or Material Barium perchlorate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
May cause fire or explosion; strong oxidizer.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep/Store away from clothing/other combustible materials.
Take any precaution to avoid mixing with combustibles.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 236-710-4
RTECSNumber SC7550000
TSCA TSCA

RUO – Research Use Only