Learn More
Barium isopropoxide, 20% w/v in isopropanol
In Sol-Gel synthesis and superconductors | CAS: 24363-37-9 | C6H14BaO2 | 255.503 g/mol
$121.81 - $575.49
Chemical Identifiers
| CAS | 24363-37-9 |
|---|---|
| Molecular Formula | C6H14BaO2 |
| Molecular Weight (g/mol) | 255.503 |
| MDL Number | MFCD00050486 |
| InChI Key | CPUJSIVIXCTVEI-UHFFFAOYSA-N |
| Synonym | barium isopropoxide, barium 2+ ; propan-2-olate, barium diisopropoxide, acmc-20alr0, barium ii isopropoxide, barium 2+ bis propan-2-olate, barium 2+ ion bis propan-2-olate |
| PubChem CID | 6101105 |
| IUPAC Name | barium(2+);propan-2-olate |
| SMILES | CC(C)[O-].CC(C)[O-].[Ba+2] |
Description
Barium isopropoxide is used in nano-materials and thin Films, catalysis synthesis, ferroelectric metals. It is also used as organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 24363-37-9 | |
| 255.503 | |
| CPUJSIVIXCTVEI-UHFFFAOYSA-N | |
| 6101105 | |
| CC(C)[O-].CC(C)[O-].[Ba+2] |
| C6H14BaO2 | |
| MFCD00050486 | |
| barium isopropoxide, barium 2+ ; propan-2-olate, barium diisopropoxide, acmc-20alr0, barium ii isopropoxide, barium 2+ bis propan-2-olate, barium 2+ ion bis propan-2-olate | |
| barium(2+);propan-2-olate |
Specifications
| 24363-37-9 | |
| 0.89 g/mL | |
| Alcohol-like | |
| Ba[OCH(CH3)2]2 | |
| 10 mL | |
| Moisture sensitive | |
| Soluble in alcohols,ketones and esters. Reacts with water. | |
| CC(C)[O-].CC(C)[O-].[Ba+2] | |
| 255.503 | |
| 255.52 | |
| Liquid |
| Yellow | |
| 22°C (72°F) | |
| C6H14BaO2 | |
| MFCD00050486 | |
| UN3274 | |
| barium isopropoxide, barium 2+ ; propan-2-olate, barium diisopropoxide, acmc-20alr0, barium ii isopropoxide, barium 2+ bis propan-2-olate, barium 2+ ion bis propan-2-olate | |
| CPUJSIVIXCTVEI-UHFFFAOYSA-N | |
| barium(2+);propan-2-olate | |
| 6101105 | |
| 20% w/v in isopropanol | |
| Barium isopropoxide |
Safety and Handling
GHS H Statement
H225-H302-H319-H336
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes serious eye irritation.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314-H335-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLATES SOLUTION, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only