missing translation for 'onlineSavingsMsg'
Learn More

Barium isopropoxide, 20% w/v in isopropanol

Catalog No. AA41338AD
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41338AD 50 mL
AA41338AB 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA41338AD Supplier Thermo Scientific Chemicals Supplier No. 041338AD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

In Sol-Gel synthesis and superconductors | CAS: 24363-37-9 | C6H14BaO2 | 255.503 g/mol

Barium isopropoxide is used in nano-materials and thin Films, catalysis synthesis, ferroelectric metals. It is also used as organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24363-37-9
Molecular Formula C6H14BaO2
Molecular Weight (g/mol) 255.503
MDL Number MFCD00050486
InChI Key CPUJSIVIXCTVEI-UHFFFAOYSA-N
Synonym barium isopropoxide, barium 2+ ; propan-2-olate, barium diisopropoxide, acmc-20alr0, barium ii isopropoxide, barium 2+ bis propan-2-olate, barium 2+ ion bis propan-2-olate
PubChem CID 6101105
IUPAC Name barium(2+);propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].[Ba+2]

Specifications

Color Yellow
Density 0.89 g/mL
Flash Point 22°C (72°F)
Odor Alcohol-like
Linear Formula Ba[OCH(CH3)2]2
Quantity 50 mL
UN Number UN3274
Sensitivity Moisture sensitive
Solubility Information Soluble in alcohols,ketones and esters. Reacts with water.
Formula Weight 255.52
Concentration or Composition (by Analyte or Components) 20% w/v in isopropanol
Physical Form Liquid
Chemical Name or Material Barium isopropoxide
Show More Show Less
Hazard Category H225-H314-H335-H336
Hazard Statement GHS H Statement
H225-H302-H319-H336
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLATES SOLUTION, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only