missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium isopropoxide, 20% w/v in isopropanol
Description
Barium isopropoxide is used in nano-materials and thin Films, catalysis synthesis, ferroelectric metals. It is also used as organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Yellow |
| Density | 0.89 g/mL |
| Flash Point | 22°C (72°F) |
| Odor | Alcohol-like |
| Linear Formula | Ba[OCH(CH3)2]2 |
| Quantity | 10 mL |
| UN Number | UN3274 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Soluble in alcohols,ketones and esters. Reacts with water. |
| Formula Weight | 255.52 |
| Show More |
Safety and Handling
| Hazard Category | H225-H314-H335-H336 |
| Hazard Statement | GHS H Statement H225-H302-H319-H336 Highly flammable liquid and vapor. Harmful if swallowed. Causes serious eye irritation. May cause drowsiness or dizziness. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLATES SOLUTION, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only