missing translation for 'onlineSavingsMsg'
Learn More

Barium iodide hydrate, 95%

Light Sensitive and Hygroscopic | CAS: 13718-50-8 | BaI2 | 391.14 g/mol

$159.93 - $413.08

Chemical Identifiers

CAS 13718-50-8
Molecular Formula BaI2
Molecular Weight (g/mol) 391.14
MDL Number MFCD00003452
InChI Key SGUXGJPBTNFBAD-UHFFFAOYSA-L
IUPAC Name barium(2+) diiodide
SMILES [I-].[I-].[Ba++]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1512614
View Documents
Thermo Scientific Chemicals
01512614
25 g
Each for $159.93
Only null left
 
AA1512622
View Documents
Thermo Scientific Chemicals
01512622
100 g
Each for $413.08
Only null left
 
Description

Description

Useful in pyrotechnic devices. A reagent for facile synthesis of poly(phenylcarbyne).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13718-50-8
391.14
SGUXGJPBTNFBAD-UHFFFAOYSA-L
[I-].[I-].[Ba++]
BaI2
MFCD00003452
barium(2+) diiodide

Specifications

White
Powder
95%
MFCD00003452
SGUXGJPBTNFBAD-UHFFFAOYSA-L
barium(2+) diiodide
391.15 (Anhydrous)
Hygroscopic; Light sensitive
13718-50-8
25 g
BaI2
Freely soluble in water. Soluble in alcohol and acetone
[I-].[I-].[Ba++]
391.14
Odorless
Barium iodide hydrate
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 237-276-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only