missing translation for 'onlineSavingsMsg'
Learn More

Barium(II) Bis(trifluoromethanesulfonyl)imide 99.0+%, TCI America™

$109.98 - $329.94

Chemical Identifiers

CAS 168106-22-7
Molecular Formula C4H2BaF12N2O8S4
Molecular Weight (g/mol) 699.614
MDL Number MFCD22200529
InChI Key VJUKAFJLZPVWME-UHFFFAOYSA-N
Synonym barium bis trifluoromethanesulfonimide, barium bis-triflimide
PubChem CID 131725281
IUPAC Name barium;1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide
SMILES C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F.[Ba]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
B51711G
View Documents
TCI America
B51711G
1 g
Each for $109.98
Only null left
 
B51715G
View Documents
TCI America
B51715G
5 g
Each for $329.94
Only null left
 
Description

Description

Specifications

Chemical Identifiers

168106-22-7
699.614
VJUKAFJLZPVWME-UHFFFAOYSA-N
131725281
C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F.[Ba]
C4H2BaF12N2O8S4
MFCD22200529
barium bis trifluoromethanesulfonimide, barium bis-triflimide
barium;1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide

Specifications

168106-22-7
C4H2BaF12N2O8S4
1 g
barium bis trifluoromethanesulfonimide, barium bis-triflimide
C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F.[Ba]
699.614
697.60
Crystalline Powder
White
MFCD22200529
1759
VJUKAFJLZPVWME-UHFFFAOYSA-N
barium;1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide
131725281
≥99.0% (T)
Barium(II) Bis(trifluoromethanesulfonyl)imide
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents