missing translation for 'onlineSavingsMsg'
Learn More

Barium hydroxide solution, Volumetric, 0.05 M Ba(OH)2 (0.1N), Solstice

Catalog No. 6002589
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002589 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6002589 Supplier Solstice Supplier No. 350501L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Volumetric, 0.05 M Ba(OH)2 (0.1N)

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

Chemical Identifiers

CAS 17194-00-2
Molecular Formula BaH2O2
Molecular Weight (g/mol) 171.341
InChI Key RQPZNWPYLFFXCP-UHFFFAOYSA-L
Synonym barium hydroxide, barium 2+ dihydroxide, bariumhydroxid, aetzbaryt, barium ii hydroxide, barium 2+ hydroxide, acmc-20alr7, barium hydroxide,anhydrous, barium hydroxide anhydrous
PubChem CID 6093286
ChEBI CHEBI:32592
IUPAC Name barium(2+);dihydroxide
SMILES [OH-].[OH-].[Ba+2]

Specifications

Physical Form Liquid
Quantity 1 L
Linear Formula Ba(OH)2
MDL Number MFCD00003443
UN Number NONH for all modes of transport
Formula Weight 171.34g/mol
Grade Volumetric
Chemical Name or Material Barium hydroxide solution