missing translation for 'onlineSavingsMsg'
Learn More

Barium hydroxide octahydrate, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥98%, Solstice

Catalog No. 6000369
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000369 500 g
6000367 100 g
6000368 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6000369 Supplier Solstice Supplier No. 31127500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Puriss. p.a., Reag. ISO, Reag. Ph. Eur.,≥98%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 12230-71-6
Molecular Formula BaH18O10
Molecular Weight (g/mol) 315.46
MDL Number MFCD00149152
InChI Key ZUDYPQRUOYEARG-UHFFFAOYSA-L
Synonym barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate
PubChem CID 17749109
IUPAC Name barium(2+);dihydroxide;octahydrate
SMILES O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]

Specifications

Physical Form Solid
Quantity 500 g
Packaging Plastic Bottle
Linear Formula Ba(OH)2 · 8H2O
UN Number 3262
Formula Weight 315.46g/mol
Percent Purity ≥98%
Grade puriss. p.a.
Chemical Name or Material Barium Hydroxide Octahydrate, Reag. Ph. Eur. reag. ISO
Hazard Statement P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H302 + H332-H314
EINECSNumber 241-234-5