missing translation for 'onlineSavingsMsg'
Learn More

Barium hydroxide octahydrate, ACS, 98+%

CAS: 12230-71-6 | BaH18O10 | 315.46 g/mol

$100.62 - $284.22

Chemical Identifiers

CAS 12230-71-6
Molecular Formula BaH18O10
Molecular Weight (g/mol) 315.46
MDL Number MFCD00149152
InChI Key ZUDYPQRUOYEARG-UHFFFAOYSA-L
Synonym barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate
PubChem CID 17749109
SMILES O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1449930
View Documents
Thermo Scientific Chemicals
01449930
250 g
Each for $100.62
Only null left
 
AA14499A1
View Documents
Thermo Scientific Chemicals
014499A1
1 kg
Each for $284.22
Only null left
 
Description

Description

Used in manufacture of alkali, glass; in synthetic rubber vulcanization, alkalizing agent in water softening, boiler scale remedy, softening water.Barium hydroxide octahydrate is used in the centromeric heterochromatin banding technique. It acts as a precursor to prepare barium titanate, barium sulfate, barium phosphate and barium sulfide. Further, it is used in analytical chemistry for the titration of weak acids especially organic acids. It is involved in the preparation of cyclopentanone, diacetone alcohol and D-Gulonic gamma-lactone. In addition to this, it finds application in ester hydrolysis such as dimethyl hendecanedioate and decarboxylation of amino acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12230-71-6
315.46
ZUDYPQRUOYEARG-UHFFFAOYSA-L
17749109
BaH18O10
MFCD00149152
barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate
O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]

Specifications

78°C
Crystalline
>98.0%
MFCD00149152
Soluble in water and methanol. Slightly soluble in ethanol. Insoluble in acetone.
O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]
315.46
315.48 (171.36 Anhydrous)
Air sensitive; Hygroscopic
Barium hydroxide octahydrate, For ACS analysis
12230-71-6
250 g
BaH18O10
barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate
ZUDYPQRUOYEARG-UHFFFAOYSA-L
barium(2+) octahydrate dihydroxide
17749109
ACS Reagent
2.18
Videos
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H332-H314

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 241-234-5

RTECSNumber : CQ9200000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only