missing translation for 'onlineSavingsMsg'
Learn More

Barium hydroxide octahydrate, ACS, 98+%

Catalog No. AA14499A1
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA14499A1 1 kg
AA1449930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA14499A1 Supplier Thermo Scientific Chemicals Supplier No. 014499A1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 12230-71-6 | BaH18O10 | 315.46 g/mol

Used in manufacture of alkali, glass; in synthetic rubber vulcanization, alkalizing agent in water softening, boiler scale remedy, softening water.Barium hydroxide octahydrate is used in the centromeric heterochromatin banding technique. It acts as a precursor to prepare barium titanate, barium sulfate, barium phosphate and barium sulfide. Further, it is used in analytical chemistry for the titration of weak acids especially organic acids. It is involved in the preparation of cyclopentanone, diacetone alcohol and D-Gulonic gamma-lactone. In addition to this, it finds application in ester hydrolysis such as dimethyl hendecanedioate and decarboxylation of amino acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12230-71-6
Molecular Formula BaH18O10
Molecular Weight (g/mol) 315.46
MDL Number MFCD00149152
InChI Key ZUDYPQRUOYEARG-UHFFFAOYSA-L
Synonym barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate
PubChem CID 17749109
SMILES O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]

Specifications

Melting Point 78°C
Physical Form Crystalline
Quantity 1 kg
Sensitivity Air sensitive; Hygroscopic
Assay Percent Range >98.0%
Density 2.18
Solubility Information Soluble in water and methanol. Slightly soluble in ethanol. Insoluble in acetone.
IUPAC Name barium(2+);dihydroxide;octahydrate
Formula Weight 315.48 (171.36 Anhydrous)
Grade ACS Reagent
Chemical Name or Material Barium hydroxide octahydrate, For ACS analysis
Show More Show Less
Hazard Category H302+H332-H314
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 241-234-5
RTECSNumber CQ9200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only