missing translation for 'onlineSavingsMsg'
Learn More

Barium hydroxide octahydrate, 99.95%, (trace metal basis)

CAS: 12230-71-6 | BaH18O10 | 315.46 g/mol

$110.39 - $312.78

Chemical Identifiers

CAS 12230-71-6
Molecular Formula BaH18O10
Molecular Weight (g/mol) 315.46
MDL Number MFCD00149152
InChI Key ZUDYPQRUOYEARG-UHFFFAOYSA-L
Synonym barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate
PubChem CID 17749109
IUPAC Name barium(2+);dihydroxide;octahydrate
SMILES O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC377820050
SDS View Documents
Thermo Scientific Chemicals
377820050
5 g Glass bottle
Each for $110.39
Only null left
 
AC377820250
SDS View Documents
Thermo Scientific Chemicals
377820250
25 g Glass bottle
Each for $312.78
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Chemical Identifiers

12230-71-6
315.46
ZUDYPQRUOYEARG-UHFFFAOYSA-L
17749109
O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]
BaH18O10
MFCD00149152
barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate
barium(2+);dihydroxide;octahydrate

Specifications

78.0°C
White
5 g
BaH18O10
MFCD00149152
barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate
ZUDYPQRUOYEARG-UHFFFAOYSA-L
barium(2+);dihydroxide;octahydrate
17749109
99.95%
Glass bottle
2.1800g/mL
12230-71-6
Powder, Crystals and/or Chunks
99.95%
Ba(OH)2·8H2O
15, 973
Solubility in water: 56g/L (15°C). Other solubilities: freely soluble in methanol, slightly soluble in ethanol, practically insoluble in acetone
O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]
315.46
315.48
Trace Metal Basis
2.18
Barium hydroxide octahydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only