missing translation for 'onlineSavingsMsg'
Learn More

Barium hydroxide octahydrate, 98+%, ACS reagent

Catalog No. AC423465000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423465000 500 g Plastic bottle
AC423461000 100 g Plastic bottle
AC423460025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC423465000 Supplier Thermo Scientific Chemicals Supplier No. 423465000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 12230-71-6 | BaH18O10 | 315.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12230-71-6
Molecular Formula BaH18O10
Molecular Weight (g/mol) 315.46
MDL Number MFCD00149152
InChI Key ZUDYPQRUOYEARG-UHFFFAOYSA-L
Synonym barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate
PubChem CID 17749109
IUPAC Name barium(2+);dihydroxide;octahydrate
SMILES O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]

Specifications

Melting Point 78.0°C
Color White
Physical Form Crystalline Flakes
Quantity 500 g
Packaging Plastic bottle
Specific Gravity 2.18
Assay Percent Range 98+%
Density 2.1800g/mL
Linear Formula Ba(OH)2·8H2O
Merck Index 15, 973
Solubility Information Solubility in water: 56g/L (15°C). Other solubilities: freely soluble in methanol, slightly soluble in ethanol, practically insoluble in acetone
Formula Weight 315.48
Percent Purity ≥98%
Grade ACS Reagent
Chemical Name or Material Barium hydroxide octahydrate, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

RUO – Research Use Only