missing translation for 'onlineSavingsMsg'
Learn More

Barium hydroxide, anhydrous, 94-98%

Catalog No. p-4407259
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1219518 50 g
AA1219530 250 g
AA12195A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1219518 Supplier Thermo Scientific Chemicals Supplier No. 01219518

CAS: 17194-00-2 | BaH2O2 | 171.341 g/mol

Cobalt(II,III) oxide is used in enamels, pigments, porcelain colorants, semiconductors, ceramics, catalysis, grinding wheels and other fields as an important inorganic material. It is a useful oxidant in chemical industry and as catalyst for organic synthesis. It is widely used as catalyst carrier and as electrode active material. It is also used in senior goggles, other filter materials, in temperature and gas sensors, in electronic ceramics, in lithium ion battery electrode materials, magnetic materials, electrochromic devices and solar energy absorbers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 17194-00-2
Molecular Formula BaH2O2
Molecular Weight (g/mol) 171.341
MDL Number MFCD00003443
InChI Key RQPZNWPYLFFXCP-UHFFFAOYSA-L
Synonym barium hydroxide, barium 2+ dihydroxide, bariumhydroxid, aetzbaryt, barium ii hydroxide, barium 2+ hydroxide, acmc-20alr7, barium hydroxide,anhydrous, barium hydroxide anhydrous
PubChem CID 6093286
ChEBI CHEBI:32592
IUPAC Name barium(2+);dihydroxide
SMILES [OH-].[OH-].[Ba+2]

Specifications

Melting Point 408°C
Physical Form Powder
Quantity 50 g
Assay Percent Range 94-98%
Solubility Information Soluble in water.
Formula Weight 171.36
Sensitivity Air sensitive; Hygroscopic
Chemical Name or Material Barium hydroxide, anhydrous
Hazard Category H302+H332-H314
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 241-234-5
RTECSNumber CQ9200000
TSCA Yes
storageNote1 Air Sensitive & Hygroscopic
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only