missing translation for 'onlineSavingsMsg'
Learn More

Barium fluoride, 99%

CAS: 7787-32-8 | BaF2 | 175.32 g/mol

Supplier:  Thermo Scientific Chemicals 012338A1

 View more versions of this product

Catalog No. AA12338A1

Only null left
Explore available promotions

Description

Description

It finds applications in ceramic flux, glass manufacturing, metallurgy, electronics and dry-film lubricants. It is transparent from ultraviolet to infrared. Barium fluoride is usually employed as a scintillator material, and useful as a transmission window material for IR spectroscopy, mostly used for analysis of fuel oil samples. Of all the optical fluorides, it is the most resistant to high-energy radiation. It is used as a preopacifying agent, and in the production of enamel, welding agents, and glazing frits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2260°C
1353°C
Colorless
1 kg
BaF2
barium fluoride, baryum fluorure french, baryum fluorure, difluorobarium, barium fluoride, ultra dry, barium ii fluoride, barium fluoride, powder, nickelous sulfate; nickel ii sulfate, barium fluoride trace metals basis
OYLGJCQECKOTOL-UHFFFAOYSA-L
barium(2+);difluoride
62670
Odorless
4.83
Barium fluoride
7787-32-8
Powder
99%
MFCD00003450
Soluble in ethanol,methanol,and aqueous solutions of ammonium chloride. Slightly soluble in water.
[F-].[F-].[Ba++]
175.32
175.36
1.4741

Chemical Identifiers

7787-32-8
175.32
OYLGJCQECKOTOL-UHFFFAOYSA-L
62670
BaF2
MFCD00003450
barium fluoride, baryum fluorure french, baryum fluorure, difluorobarium, barium fluoride, ultra dry, barium ii fluoride, barium fluoride, powder, nickelous sulfate; nickel ii sulfate, barium fluoride trace metals basis
[F-].[F-].[Ba++]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P301+P310-P304+P340-P312-P330-P501c

H301-H332

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 232-108-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only