missing translation for 'onlineSavingsMsg'
Learn More

Barium fluoride, 99.9% (metals basis)

Catalog No. AA3638022
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3638022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3638022 Supplier Thermo Scientific Chemicals Supplier No. 03638022

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7787-32-8 | BaF2 | 175.32 g/mol

Barium fluoride is used as a material to make optical components such as lenses, in windows for infrared spectroscopy, in particular in the field of fuel oil analysis, used as a preopacifying agent and in enamel and glazing frits production. It is also used in the production of welding agents, metallurgy, as a molten bath for refining aluminum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7787-32-8
Molecular Formula BaF2
Molecular Weight (g/mol) 175.32
MDL Number MFCD00003450
InChI Key OYLGJCQECKOTOL-UHFFFAOYSA-L
Synonym barium fluoride, baryum fluorure french, baryum fluorure, difluorobarium, barium fluoride, ultra dry, barium ii fluoride, barium fluoride, powder, nickelous sulfate; nickel ii sulfate, barium fluoride trace metals basis
PubChem CID 62670
SMILES [F-].[F-].[Ba++]

Specifications

Boiling Point 2260°C
Melting Point 1353°C
Color Colorless
Physical Form Fused Lump
Quantity 100 g
Assay Percent Range 99.99% (Metals basis)
Solubility Information Soluble in methanol,ethanol,water 1.2g/L.
IUPAC Name barium(2+);difluoride
Formula Weight 175.36
Odor Odorless
Refractive Index 1.4741
Density 4.83
Chemical Name or Material Barium fluoride
Show More Show Less
Hazard Category H301-H332
Precautionary Statement P261-P264b-P270-P271-P301+P310-P304+P340-P312-P330-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 232-108-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only