Learn More
Barium dodecairon nonadecaoxide
CAS: 12047-11-9 | BaFe12O19
$187.12 - $670.82
Chemical Identifiers
| CAS | 12047-11-9 |
|---|---|
| Molecular Formula | BaFe12O19 |
| MDL Number | MFCD00075637 |
| Synonym | Barium ferrite |
Description
Barium dodecairon nonadecaoxide is used in subwoofer magnets to provide power. It is also used in new high tech magnetic tapes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 12047-11-9 | |
| BaFe12O19 | |
| MFCD00075637 | |
| Barium ferrite |
Specifications
| >1315°C | |
| 12047-11-9 | |
| 25 g | |
| MFCD00075637 | |
| Barium ferrite | |
| 1111.48 |
| Black | |
| Powder | |
| BaFe12O19 | |
| UN1564 | |
| Insoluble in water. | |
| Barium dodecairon nonadecaoxide |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
EINECSNumber : 234-974-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only