missing translation for 'onlineSavingsMsg'
Learn More

Barium Diphenylamine Sulfonate, For Chlorine, Chlorine Dioxide, Certified, 0.1% (w/v), LabChem™

Numéro de catalogue. LC116307
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
LC116307 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. LC116307 Fournisseur LabChem Code fournisseur LC116307
Il en reste null

Spécifications

Color Clear, colorless
Concentration 0.1% (w/v)
Description CAS No.: 6211-24-1
Format Liquid
Packaging Poly Bottle
Quantity 125 mL
Storage Requirements Room Temperature
Type Barium Diphenylamine Sulfonate