missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Chloride, Dihydrate, Purified, 95%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 500 g |
| Percent Purity | 95% |
| Grade | Purified |
| Packaging | Poly Bottle |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.